Macro Cuvette with PTFE Cover and Magnetic Stirrer Slot (Lightpath: 10 mm), Type 1MS
Fournisseur: FIREFLYSCI, INC.
Certificats
À propos de cet article
A Type 1 standard spectrophotometer cuvette that comes with a circular groove, which fits a P73 Magnetic Stirrer. The P73 is agitated by a P82 magnetic cuvette stirrer and the rotating P73 creates a vortex, mixing the solution while not interfering with measurements.
- Fire-fused to ensure that cuvettes are robust and durable
- Available in different materials: Optical Glass, UV Quartz, IR Quartz
- Available in 10 mm lightpath
- Two windows optically polished
- Includes PTFE cover
Features circular channel at the bottom in the cell that is 3mm deep and 7mm wide, which fits a standard P73 macro cuvette stirrer. Includes P73 Stirrer for 10 mm cells.
Frequently Bought Together