SHURMount™ Plus Liquid Coverslip
Catalog # CA76632-838
Supplier: General Data Healthcare
Specifications
- Volume:4 oz.
- Description:SHURMount™ Plus liquid coverslip
- Material:Liquid
- Cat. no.:CA76632-838
Specifications
About this item
Spreads thinly and evenly to form a permanent, scratch-resistant layer that acts as a coverslip or can be used in conjunction with a coverslip.
- Toluene based, compatible with most applications
- Minimizes the need to purchase coverslips
Acrylic resin allows rapid drying for hand coverslipping.