Eisco Serological Pipettes, Class B
Eisco Serological Pipettes, Class B
Supplier:   Wards
Eisco Serological Pipettes, Class B
Supplier:  Wards
Certificates
Eisco Serological Pipettes, Class B
Eisco Serological Pipettes, Class B
4 Options Available Below
Product Details & Documents
Class B color-coded Serological Pipettes made of high quality heavy duty borosilicate 3.3 glass with blue graduations.
  • Class B, conforms with DIN IS 835 standards
  • Calibrated for delivery to jet
  • Last drop expelled by blowing
  • Blue graduations for optimal contrast against clear liquids
  • Color coded for easy identification
  • Reusable
Class B tolerance, ±0.200 ml. Autoclavable, with superior chemical durability. Low temperature gradient - can withstand high temperatures and thermal shocks.
Eisco Labs Borosilicate glass has the following heat tolerances:
- Strain point 515 °C
- Annealing point 565 °C
- Softening point 820 °C
Product Family Options
Product Information
  • Capacité
    Graduation
    Tolerance
    Color Code
    Availability
    Price
  • 1 ml
    0.01 ml
    ±0.010 ml
    Yellow
    Log in for availability
    Log in for availability
    Each
    Each - 9,55 $
    9,55 $
    Each
       9,55 $
    Supplier Number: CH0572D
  • 5 ml
    0.05 ml
    ±0.050 ml
    Red
    Log in for availability
    Log in for availability
    Each
    Each - 10,35 $
    10,35 $
    Each
       10,35 $
    Supplier Number: CH0572F
  • 10 ml
    0.10 ml
    ±0.100 ml
    Orange
    Log in for availability
    Log in for availability
    Each
    Each - 11,80 $
    11,80 $
    Each
       11,80 $
    Supplier Number: CH0572G
  • 25 ml
    0.20 ml
    ±0.200 ml
    White
    Log in for availability
    Log in for availability
    Each
    Each - 12,20 $
    12,20 $
    Each
       12,20 $
    Supplier Number: CH0572H