Microscope Slide Mailers, Holds 5 Microscope Slides, Translucent, 100/Box
Product Details & Documents
Translucent mailer for microscope slides.
- Snap top closure
- 100/box
Slide mailers are designed to hold five (5) 1×3" standard microscope slides.
Spécifications
- Color:Translucent
- Désignation:Slide Mailer for 5 Slides