Control Slides IHC 5-MC
Marketsource
 :   Mopec
Control Slides IHC 5-MC
 :  Mopec
Marketsource
Control Slides IHC 5-MC
Control Slides IHC 5-MC
All control slides are on Mopec IHC Enhanced Hydrophilic Adhesive Slides using Formalin Fixed Paraffin Embedded (FFPE) human tissue.
  • Product specific reactivity is guaranteed for one year from date of receipt
  • Revalidate after one year to verify continued reactivity
  • All controls are cut in serial sections at 4 microns
  • Standard tissue placement is mid-high on slide to facilitate placement of patient and control tissue on the same slide
  • All slides are air dried
Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 60 to 80 °F in a humidity controlled and light-deprived environment.
 : Case of 25 contains 24 unstained slides and 1 stained slide. Case of 100 contains 98 unstained slides and 2 stained slides.
Product Family Options
  • Désignation
  • Mopec control slides IHC 5-MC
     : SC002-25
    Marketsource
    Marketsource
  • Mopec control slides IHC 5-MC
    Case of 100
    Case of 100 - $2,591.32
    $2,591.32
    Case of 100
       $2,591.32
     : SC002-100