APM Young Body Bag, 8 Mil PVC, Center Zipper, White
Fournisseur: Mortech Manufacturing
Certificats
À propos de cet article
Mopec in partnership with American Professional Manufacturing provide excellent body bags.
- Custom-made, lockable slider for easy opening and closing
- Dielectrically sealed seams
- Center zipper
- Color: white
- Leak resistance
Materials: 8 mil PVC
Dimensions: Various sizes
Frequently Bought Together