ContactUsLinkComponent
ProLite™ His-Tag Protein Gel Staining Kit, AAT Bioquest
Catalog # 76779-082
Supplier:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
ProLite™ His-Tag Protein Gel Staining Kit, AAT Bioquest
Catalog # 76779-082
Supplier:  AAT BIOQUEST INC
Supplier Number:  18010
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Pack type:
    Plastic vial
  • Description:
    ProLite™ His-Tag Protein Gel Staining Kit "Green Fluorescence"
  • Size:
    10 gels
  • Cat. no.:
    76779-082
Specifications
About this item
This kit requires very little hands-on time, allowing for rapid protein expression screening in a variety of gel types using an imaging equipment with a standard FITC filter.
  • Green fluorescence
  • Compatible with standard FITC filter
Kit provides a fast, sensitive, and highly specific fluorescent stain for visualizing His-tagged fusion proteins directly in a polyacrylamide gel following electrophoresis
It detects nanogram of His-tagged proteins directly in gels, thus eliminating the additional western blotting step.
Caution: For research use only.