ContactUsLinkComponent
Portelite™ Fluorimetric Total Nucleic Acid Quantitation Kit
Catalog # 76779-116
Supplier:  AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Portelite™ Fluorimetric Total Nucleic Acid Quantitation Kit
Catalog # 76779-116
Supplier:  AAT Bioquest
Supplier Number:  17631
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Specifications
  • Description:
    Kit is designed to rapidly measure the total amounts of nucleic acids, including double-stranded DNA (dsDNA), single-stranded DNA (ssDNA) and RNA in a sample
  • Size:
    200 tests
  • Cat. no.:
    76779-116
Specifications
About this item
Portelite™ fluorimetric total nucleic acid quantitation kit is optimized for measuring the total amounts of nucleic acids with CytoCite™ or Qubit® fluorometers.
  • Optimized for cytocite and qubit fluorometers
  • Sensitive and indiscriminate binding
The kit has all the essential reagents, including Helixyte™ Green ssDNA reagent, dilution buffer, and pre-diluted DNA standards.
Packaging: Plastic vial.
Caution: For research use only.