Helixyte™ Green Fluorimetric Total Nucleic Acid Quantitation Kit
76779-040
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
:
- Description:Kit is designed to measure total amounts of nucleic acids, including double-stranded DNA (dsDNA), single-stranded DNA (ssDNA) and RNA in an easy and accurate format
- Size:200 tests
- Cat. no.:76779-040
Helixyte™ Green fluorimetric total nucleic acid quantitation kit is optimized for measuring the total amounts of nucleic acids with a fluorescence microplate reader.
- Optimized for microplate readers
- Sensitive and indiscriminate binding
The kit has all the essential reagents, including Helixyte™ Green all reagent, dilution buffer, and pre-diluted DNA standards.
: Plastic vial.
: For research use only.