ReadiScript™ RT Reverse Transcription Kit | Avantor
ReadiScript™ RT Reverse Transcription Kit
Convenient tool for cDNA synthesis used in gene expression analysis with qPCR, 50 reactions
Catalog # 76779-098
Supplier:   AAT Bioquest
Fournisseur #:  60100
ReadiScript™ RT Reverse Transcription Kit
Catalog # 76779-098
Supplier:  AAT Bioquest
CAS Number:  
ReadiScript™ RT Reverse Transcription Kit
ReadiScript™ RT Reverse Transcription Kit
Convenient tool for cDNA synthesis used in gene expression analysis with qPCR, 50 reactions
CAS Number:  
Price
$869.10
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
The two-tube kit format helps to set up the reaction faster and is optimized to yield cDNA over a broad dynamic range.
  • Robust protocol
  • Broad dynamic range
  • Useful for gene expression analysis
The 5X supermix contains a unique blend of oligo(dT) and random primer mix to prevent the 5' and 3' bias of the target genes. The reaction mix contains a potent blend of RNase inhibitors to protect RNA during the setup and reverse transcription process.
The resulting cDNA product is compatible with all qPCR chemistries or conventional end-point RT-PCR.
 : Plastic vial.
 : For research use only.
Spécifications
Spécifications
  • Désignation:
    Convenient tool for cDNA synthesis used in gene expression analysis with qPCR
  • Taille:
    50 reactions