ReadiLink™ iFluor® 555 Oligo and ssDNA Labeling Kit
Supplier: AAT Bioquest
Certificates
About this item
The kit is optimized for efficient labelling and contains all the essential reagents required for efficient labelling of ssDNA or oligos.
- Robust protocol
- Bright labeling
- Photostable fluorescence
The resulting iFluor® 555-labelled DNA probes are ideally suited for biological applications, e.g., electrophoretic mobility shift assays (EMSA), Northern and Southern blots, colony or in situ hybridizations.
Packaging: Packaged in plastic vials.
Caution: For research use only.
Frequently Bought Together