PIP® Mission™ Indirect Vent Goggles
76944-638
: PIP
- Frame Color:Green
- Description:Mission™ Indirect vent goggles
- Cat. no.:76944-638
- Lens Coating:Anti-fog (FogLess®) 3Sixty™
- Lens Tint:Clear
- Lens Material:PC
Indirect vent goggles feature a green body, clear lens with FogLess® 3Sixty™ coating, and a Neoprene strap.
- Indirect vent for liquid splash protection
- Clear polycarbonate lens with anti-scratch and FogLess® 3Sixty™ coating
- Adjustable neoprene strap provides a comfortable fit
- Strap buckle for easy donning when wearing head protection
- Fits over most prescription glasses
- Comfortable TPR seal for comfort and extended use
Lightweight, semi-rimless design provides all-day user comfort. The sleek, sporty style with wrap-around lens protects the wearer against projectiles, impact, and liquid splash. FogLess® 3Sixty™ provides the ultimate anti-fog protection.
: ANSI Z87.1-2015 D3 Splash/Droplet compliant.