PIP® Anser™ Semi-Rimless Safety Glasses
76944-628
: PIP
- Frame Color:Black
- Description:Anser™ Semi-rimless safety glasses
- Cat. no.:76944-628
- Lens Tint:Clear
Semi-rimless safety glasses feature a black frame, clear lens, and FogLess® 3Sixty™ coating.
- Lightweight, semi-rimless design provides all day user comfort
- Bayonet temples with flexible rubber tips offer a comfortable, secure fit
- Soft rubber nose pads help keep the glasses in place
- Supplied with an adjustable neck cord
- 9 Base curve lens
Sleek, sporty style with wrap-around lens protection. The FogLess® 3Sixty™ enhanced hydrophobic coating resists lens fogging and provides clear vision in extreme working conditions.
: Meets ANSI Z87.1+.