PIP® Recon™ Rimless Safety Glasses
Catalog # 76944-626
Supplier: PIP
Specifications
- Frame Color:Clear
- Description:Recon™ rimless safety glasses
- Cat. no.:76944-626
- Lens Tint:Clear
Specifications
About this item
Rimless safety glasses feature a clear temple, clear lens, and anti-scratch FogLess® 3Sixty™ coating.
- Lightweight, rimless design provides all day comfort
- Flexible bayonet temples contain rubber pads for a secure, non-slip fit
- Adjustable rubber nosepiece provides a comfortable fit
- 9 Base curve lens
Lightweight, rimless design provides an ergonomic fit, with flexible bayonet temples that contain rubber pads for an improved fit. The FogLess® 3Sixty™ enhanced hydrophobic coating resists lens fogging and provides clear vision in extreme working conditions.
Certifications: Meets ANSI Z87.1+. Meets CSA Z94.3.