Assurance® Advanced Chemical Protection 25 mil Butyl Rubber Gloves, Rough Grip
: Avantor
25 mil butyl rubber gloves with a rough grip, available in 14" length.
- Rough grip
- Ergonomic hand design
- Easy to decontaminate
- Excellent holdout
- Made in the USA
Gloves protect against hazardous chemicals and materials.
Frequently Bought Together