VWR® One-Step RT-qPCR Kit
VWR® One-Step RT-qPCR Kit
Supplier:  VWR International
Certificates
VWR® One-Step RT-qPCR Kit
1 Options Available Below
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
VWR® One-Step RT-qPCR Kit is designed to facilitate rapid, sensitive, and precise detection and quantification of various RNA sequences via dye-based qPCR. It features a distinctive universal passive reference dye that is compatible with ROX-dependent and ROX-independent qPCR instruments.
  • For the detection and quantification of RNA targets using the SYBR®/FAM channel
  • Ready-to-use kit containing all necessary reaction components, except for primers and RNA template
  • Suitable for a variety of RNA sample types
  • Includes Hot Start Taq DNA Polymerase and VWR® Reverse Transcriptase for improved reaction specificity
  • Incorporates dUTP for carryover contamination prevention
  • VWR® One-Step Reaction Mix is supplied at a 2X concentration
The VWR® RT Enzyme Mix, supplied at a 20X concentration, contains the specially designed, temperature-controlled VWR® Reverse Transcriptase and Murine RNase Inhibitor to assist in preventing RNA degradation. This temperature-controlled RT enzyme prevents unwanted non-specific activity and displays superior thermostability in comparison to other reverse transcriptases.
Caution: Store at -20°C. For research use only.
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • 73212-0100
  • 73212-0500
  • 73212-1000
  • 73212-2500
  • Application:
  • RT-qPCR dye
  • RT-qPCR dye
  • RT-qPCR dye
  • RT-qPCR dye
  • One step:
  • Yes
  • Yes
  • Yes
  • Yes
Specifications
Product Family Options
Product Information
  • Désignation
    No. of Reactions
    Availability
    Price
  • VWR® One-Step RT-qPCR kit
    100
    Discontinued
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    73212-0100
    Application
    RT-qPCR dye
    One step
    Yes
  • VWR® One-Step RT-qPCR kit
    500
    Discontinued
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    73212-0500
    Application
    RT-qPCR dye
    One step
    Yes
  • VWR® One-Step RT-qPCR kit
    1000 (2x 500)
    Log in for availability
    Log in for availability
    Each (1,000Assays)
    Each (1,000Assays) - $1,568.86
    $1,568.86
    Each (1,000Assays)
       $1,568.86
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    73212-1000
    Application
    RT-qPCR dye
    One step
    Yes
  • VWR® One-Step RT-qPCR kit
    2500 (5× 500)
    Discontinued
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    73212-2500
    Application
    RT-qPCR dye
    One step