Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Introduction to materials science
- Cat. no.:470336-176
- Includes:SDS and instructions
- No. of Uses:15
Specifications
About this item
Explore the four main types of materials: metals, polymers, ceramics, and composites.
- Test the physical and mechanical properties of different metal samples
In this activity, students will be introduced to the field of materials science as they explore the four main types of materials: metals, polymers, ceramics, and composites. They will test the physical and mechanical properties of different material samples, then categorize each material based on their results. Kit contains enough material for 15 groups. Teacher’s Guide and Student Study Guide copymasters included.