Wards® DNA Extraction Lab Activity
Product Details & Documents
Use biological detergents, enzymes, and ethanol to isolate DNA from plant material.
- Teacher's manual and study guide are included
In this lab you will learn the history of the discovery of DNA and DNA structure. Understand the nature of genetic inheritance and the role of DNA and proteins in genetic expression while using biological detergents, enzymes, and ethanol to isolate DNA from plant material. You need to supply the plant material. Kit contains enough materials for 15 groups. Teacher’s Manual and Student Study Guide copymasters are included.
Delivery information: Includes 250 ml 7.5% SDS/1.5% NaCl; 20 ml Ethanol, 95%; 1 Pepsin (to make 25 ml 0.5% solution); 10 Zipper bags; 5 Filters; 5 Plastic tubes; 10 Graduated pipettes; 5 Stirrers.
Spécifications
- Désignation:DNA extraction lab activity
- Includes:SDS and instructions
- No. of Uses:5