Eisco Plastic Scoops
Supplier: Wards
Jump to:
About this item
Pack of 12 or 6 weighing scoops with labeled handles and flat bottoms.
- Made of heat- and chemical-resistant polypropylene plastic
- Scoop handles are labeled with capacity for easy identification
- Flat bottom design allows scoops to rest upright for weighing and filling
Autoclavable and dishwasher safe for easy cleaning.
8 Options Available Below
Product Details & Documents
Pack of 12 or 6 weighing scoops with labeled handles and flat bottoms.
- Made of heat- and chemical-resistant polypropylene plastic
- Scoop handles are labeled with capacity for easy identification
- Flat bottom design allows scoops to rest upright for weighing and filling
Autoclavable and dishwasher safe for easy cleaning.
Product Family Options
Product Information
- DésignationCapacitéAvailabilityPrice
- Plastic scoops2 mlDiscontinued-Supplier Number: -
- Supplier Number: CH0461APK12
- Plastic scoops5 mlDiscontinued-Supplier Number: -
- Supplier Number: CH0461BPK12
- Plastic scoops10 mlDiscontinued-Supplier Number: -
- Supplier Number: CH0461CPK12
- Plastic scoops50 mlDiscontinued-Supplier Number: -
- Supplier Number: CH0461EPK12
- Plastic scoops100 mlDiscontinued-Supplier Number: -
- Supplier Number: CH0461FPK6
- Plastic scoops250 mlDiscontinued-Supplier Number: -
- Supplier Number: CH0461GPK6
- Plastic scoops500 mlDiscontinued-Supplier Number: -
- Supplier Number: CH0461HPK6
- Plastic scoops1000 mlDiscontinued-Supplier Number: -
- Supplier Number: CH0461IPK6