OPTI-MAX® Replacement Check Valves | Avantor
OPTI-MAX® Replacement Check Valves
Supplier:   Optimize
OPTI-MAX® Replacement Check Valves
Supplier:  Optimize
Certificates
OPTI-MAX® Replacement Check Valves
OPTI-MAX® Replacement Check Valves
13 Options Available Below
Product Details & Documents
Each ball and seat set is individually matched to ensure a lock-and-key fit. This guarantees three bubbles/minute or less in the industry standard nitrogen bubble test; common acceptable standards are 12 bubbles or less. We have also minimized the ball travel distance within the check valve; less travel distance equals quicker seating and more accurate pump metering. Simply put, tighter specifications make a better valve.
  • PEEK, PPS, and/or stainless steel body with ruby, sapphire, and/or ceramic ball and seat
  • Ball and seat set is individually matched
  • Can function in either inlet or outlet position
  • Interchangeable across multiple pump brands
A single OPTI-MAX® cartridge is interchangeable across multiple pump brands, and will function in either the inlet or outlet position. The same replacement cartridge will fit into OPTI-MAX® housings for pumps made by waters, shimadzu, agilent/HP, and most other OEM solvent delivery systems. This allows you to minimize the number of spare check valve cartridges you keep on hand, and dramatically reduce the cost of inventory required to cover your maintenance needs.
Delivery information: Includes two replacement check valve cartridges.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    For Use With
    Availability
    Price
  • OPTI-MAX® Standard ¹/₈" Ø replacement cartridge, ruby ball/sapphire seat
    PEEK
    Fits xx-49-xxxxx & xx-59-xxxxx OPTI-MAX housings
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 2
    Pack of 2 - $1,024.65
    $1,024.65
    Pack of 2
       $1,024.65
    Supplier Number: 10-59-02004
  • OPTI-MAX® Standard ¹/₈" Ø replacement cartridge, ruby ball/sapphire seat
    Stainless steel
    Fits xx-49-xxxxx & xx-59-xxxxx OPTI-MAX housings
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 2
    Pack of 2 - $1,037.57
    $1,037.57
    Pack of 2
       $1,037.57
    Supplier Number: 10-57-02004
  • OPTI-MAX® Standard ¹/₈" Ø replacement cartridge, ceramic ball and seat
    PEEK
    Dionex DQP, GPM-1, GPM-2, DXP, AGP (analytical), DRP, 2000, 4000, GP40/IP20
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 2
    Pack of 2 - $1,028.41
    $1,028.41
    Pack of 2
       $1,028.41
    Supplier Number: 10-58-02004
  • OPTI-MAX® Standard ¹/₈" Ø replacement cartridge, ceramic ball and seat
    Stainless steel
    Waters® 225 µl head (#3) (510, 590, 600/600E, 610, 650, 6K/6KA)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 2
    Pack of 2 - $1,037.95
    $1,037.95
    Pack of 2
       $1,037.95
    Supplier Number: 10-56-02004
  • OPTI-MAX® EXP® Double ball, ¹/₁₆" replacement cartridge, ceramic ball and seat
    SS Cart/PEEK endcaps
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 2
    Pack of 2 - $1,985.82
    $1,985.82
    Pack of 2
       $1,985.82
    Supplier Number: 10-86-02004
  • OPTI-MAX® EXP® Single Ball Upgrade, ¹/₁₆" replacement cartridge, ceramic ball and seat
    SS Cart/PEEK endcaps
    SSI ultra CP (For OEM and standard OPTI-MAX housings)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 2
    Pack of 2 - $1,244.47
    $1,244.47
    Pack of 2
       $1,244.47
    La quantité minimale de commande est de 1
    Supplier Number: 10-76-02004
  • OPTI-MAX® Microbore ¹/₁₆" Ø replacement cartridge, ceramic ball and seat
    Stainless steel
    Waters® 2690/alliance, varian 616
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 2
    Pack of 2 - $1,145.34
    $1,145.34
    Pack of 2
       $1,145.34
    Supplier Number: 10-46-02004
  • OPTI-MAX® Microbore ¹/₁₆" Ø replacement cartridge, ceramic ball and seat
    PEEK
    Waters® 625/626
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 2
    Pack of 2 - $1,134.10
    $1,134.10
    Pack of 2
       $1,134.10
    Supplier Number: 10-48-02004
  • OPTI-MAX® Microbore ¹/₁₆" Ø replacement cartridge, ruby ball/sapphire seat
    Stainless steel
    Fits xx-49-xxxxx & xx-59-xxxxx OPTI-MAX housings
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 2
    Pack of 2 - $1,134.10
    $1,134.10
    Pack of 2
       $1,134.10
    Supplier Number: 10-47-02004
  • OPTI-MAX® Microbore ¹/₁₆" Ø replacement cartridge, ruby ball/sapphire seat
    PEEK
    Fits xx-49-xxxxx & xx-59-xxxxx OPTI-MAX housings
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 2
    Pack of 2 - $1,129.02
    $1,129.02
    Pack of 2
       $1,129.02
    Supplier Number: 10-49-02004
  • OPTI-MAX® ¹/₃₂" Ø Replacement cartridge, ruby ball/ceramic seat
    PEEK
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 2
    Pack of 2 - $1,544.67
    $1,544.67
    Pack of 2
       $1,544.67
    Supplier Number: 10-39-02004
  • OPTI-MAX® Extended Flow ³/₁₆" replacement cartridge, ceramic ball and seat
    Stainless steel
    Waters® 400 µl (#4) and 900 µl (#6) heads (delta prep 3000, 600/600E, 650)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 2
    Pack of 2 - $1,192.65
    $1,192.65
    Pack of 2
       $1,192.65
    Supplier Number: 10-66-02005
  • OPTI-MAX® Extended Flow ³/₁₆" replacement cartridge, ruby ball/sapphire seat
    Stainless steel
    Fits xx-49-xxxxx & xx-59-xxxxx OPTI-MAX housings
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 2
    Pack of 2 - $1,188.95
    $1,188.95
    Pack of 2
       $1,188.95
    Supplier Number: 10-67-02005