BioX Cryogenic Vials, Barcoded, Racked, SBS Format
BioX Cryogenic Vials, Barcoded, Racked, SBS Format
Supplier:   TWD Scientific
BioX Cryogenic Vials, Barcoded, Racked, SBS Format
Supplier:  TWD Scientific
Certificates
BioX Cryogenic Vials, Barcoded, Racked, SBS Format
BioX Cryogenic Vials, Barcoded, Racked, SBS Format
4 Options Available Below
Product Details & Documents
Barcoded cryovials racked in SBS format 96 well racks.
  • 2D Barcodes
  • Automation friendly
  • Internal thread
This barcoded tube/rack combo is the ideal off-the-shelf option for automated high throughput screening applications.
Product Family Options
Informations sur le produit
  • Désignation
    Capacité
    Cap Color
    Sterility
    Availability
    Price
  • SBS format minitube, barcoded 2D, racked
    0.25 ml
    Blue
    Sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,920
    Case of 1,920 - $2,594.59
    $2,594.59
    Case of 1,920
       $2,594.59
    Minimum order quantity is 1
    Supplier Number: BX89-4233
  • SBS format minitube, barcoded 2D, racked
    0.5 ml
    Blue
    Sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,920
    Case of 1,920 - $2,594.59
    $2,594.59
    Case of 1,920
       $2,594.59
    Minimum order quantity is 1
    Supplier Number: BX89-4253
  • SBS format minitube, barcoded 2D, racked
    0.75 ml
    Blue
    Sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,920
    Case of 1,920 - $2,594.59
    $2,594.59
    Case of 1,920
       $2,594.59
    Minimum order quantity is 1
    Supplier Number: BX89-4283
  • SBS format minitube, barcoded 2D, racked
    1.0 ml
    Blue
    Sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,920
    Case of 1,920 - $2,594.59
    $2,594.59
    Case of 1,920
       $2,594.59
    Minimum order quantity is 1
    Supplier Number: BX89-4213