Order Entry
Canada
ContactUsLinkComponent
Anti- Mouse Monoclonal Antibody [clone: 2C4]
Catalog # 77007-496
Supplier:  Genscript
Anti- Mouse Monoclonal Antibody [clone: 2C4]
Catalog # 77007-496
Supplier:  Genscript
Supplier Number:  A02159-40

Specifications

  • Clonality:
    Monoclonal
  • Clone:
    2C4
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Multiple
  • Size:
    40 μg
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Storage buffer:
    Lyophilized with PBS, pH 7.2, containing 0.02% sodium azide
  • Storage temperature:
    The lyophilized product remains stable up to 1 year at −20 °C from date of receipt. Upon reconstitution, it can be stored for 2 - 3 weeks at 2...8 °C or for up to 12 months at −20 °C or below. Avoid repeated freezing and thawing cycles
  • Concentration:
    Reconstitute the lyophilized antibody with deionized water (or equivalent) to a final concentration of 0.5 mg/ml.
  • Immunogen:
    AAV2 capsids
  • Cat. no.:
    77007-496
  • Packaging:
    Vial

Specifications

About this item

Adeno-associated virus type 2 (AAV2) is a valuable vector for gene therapy. It is one member of the human parvovirus family which depends on co-infection with helper viruses such as adenovirus or herpes virus for efficient reproduction. The AAV2 capsid (intact particle) comprises three types of subunits, VP1, VP2, and VP3, a total of 60 copies in a ratio of 1:1:8 (VP1:VP2:VP3). For the application of AAV2 vector in gene therapy, the determination of total capsid titer is essential, accurate characterization and quantification of purified AAV2 particle product represent a critical step for clinical application. GenScript Anti-AAV2 (intact particle) Antibody (2C4), mAb, Mouse is produced from cell culture in vitro under conditions free from animal-derived components.

  • High specificity
  • High affinity
  • High stability

GenScript’s AAV antibody is a useful tool for evaluation of virus expression level, quantification of virus titer, identification of capsid proteins in AAV production, and analysis of AAV immunogenicity.

Type:
Antigen:
Clonality: Monoclonal
Clone: 2C4
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Multiple