A1CNow®+ Systems 10 Test Kit | Avantor
A1CNow®+ Systems 10 Test Kit
18...28 °C (64...82 °F) for a cumulative total of 4 months
Catalog # 76994-116
Supplier:   PTS Diagnostics
Fournisseur #:  3024
A1CNow®+ Systems 10 Test Kit
Catalog # 76994-116
Supplier:  PTS Diagnostics
CAS Number:  
A1CNow®+ Systems 10 Test Kit
A1CNow®+ Systems 10 Test Kit
18...28 °C (64...82 °F) for a cumulative total of 4 months
CAS Number:  
Price
$317.34
Log in pour la disponibilité
Each
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
The A1CNow®+ system provides healthcare professionals with a fast and easy way of obtaining accurate A1C results with just a fingerstick.
  • More efficient than the lab
  • Results in 5 minutes
  • Small (5 µl) blood sample
  • Portable – use in multiple exam rooms
  • Easy to use – minimal training required
  • No maintenance
  • Available with IFCC (mmol/mol), Measuring ranges; 4 to 13% A1C
A1CNow+ test system provides quantitative measurement of the percent of glycated hemoglobin (% A1C) levels in capillary fingerstick or venous whole blood samples. The test is for professional use to monitor glycemic control in people with diabetes. The test takes only 5 minutes and tells you how well the patient has been controlling their blood sugar over the past 2 to 3 months.
A1CNOW®+ LAB QUALITY RESULTS AT THE POINT OF CARE. The A1CNow®+ system provides healthcare professionals with a fast and easy way of obtaining accurate A1C results with just a fingerstick. This innovative technology enables clinicians to communicate face-to-face with patients about their diabetes control in minutes, not days. The analyzer, test cartridges, and shaker kit pouches may be used until the expiration date printed on the box when stored refrigerated at 2 to 8 °C (36 to 46 °F).
Certifications: NGSP Certified, CLIA-waived, IFCC-traceable, FDA-cleared, CE-marked.
Caution: Note: Product can go in and out of refrigeration as long as it does not spend more than a total of 4 months outside of refrigeration.
Spécifications
Spécifications
  • Température de stockage:
    18...28 °C (64...82 °F) for a cumulative total of 4 months