SteraMist Surface Unit
Version 103 110V SteraMist Surface Unit
Catalog # 76919-438
Supplier: TOMI ENVIRONMENTAL SOLUTIONS, INC
Fournisseur #: SUR-103
Jump to:
Product Details & Documents
The SteraMist Surface Unit is a portable, handheld point-and-spray disinfection and decontamination system.
- Mobile for rapid deployment throughout a variety of spaces
- Economical, non-corrosive, and easy to apply
- Easily incorporated into daily cleaning protocols
- Portable and rugged design for easy storage
- 30’ removable applicator cord helps you reach a wide radius
- Routine disinfection for large areas and surfaces
The SteraMist Surface Unit is the perfect solution for facility surface disinfection, bringing you handheld control with more movement and mobility around the treated area.
Delivery information: Includes SteraMist handheld applicator, tool kit and 1 gal. DI water cartridge
Spécifications
- Désignation:Version 103 110V SteraMist Surface Unit
Services
Informations sur le produit
- Catalog #DescriptionAvailabilityPrice
- 76919-416Additional one-year warranty for purchased SteraMist products and accessoriesLog in pour la disponibilitéEach - $5,640.60
- 76919-424Corrective calibration to ensure the SteraMist Surface Unit is running on proper levels, anually recommendedLog in pour la disponibilitéEach - $684.93
- 76919-422Repair work to maintain your SteraMist equipment, quoted at time of receipt of unit and after evaluationLog in pour la disponibilitéEach - $1,611.60
- 76919-432Training and continued support on SteraMist equipment, mandatory for first time users (i.e. protocol and policy development, troubleshooting, assistance as-needed etc.)Log in pour la disponibilitéEach - $10,475.40
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Replacement SteraMist applicator, for SteraMist Surface Unit and SteraMist Environment SystemReplacement SteraMist applicator, for SteraMist Surface Unit and SteraMist Environment System76919-436Log in pour la disponibilitéEach - $8,676.96
Informations sur le produit
- Catalog #AvailabilityPrice
- Stainless steel cart (only)