SteraMist BIT Solution
SteraMist BIT Solution
SteraMist BIT Solution
Certificates
SteraMist BIT Solution
SteraMist BIT Solution
5 Options Available Below
Product Details & Documents
  • All SteraMist products exclusively use low-percentage 7.8% Hydrogen Peroxide as a sole active ingredient
  • Each SteraMist product utilizes the same BIT™ Solution blend for a wide array of applications
  • BIT Solution kills within seconds of contact
  • Non-corrosive to enure superior surface compatibility
  • No toxic byproducts
  • No mixing or dilution
  • No wiping or rinsing
Documents
Product Family Options
Product Information
  • Désignation
    Taille
    Volume
    Weight
    Availability
    Price
  • SteraMist BIT Solution for use with all SteraMist Systems
    14x14x12"
    32 fl. Oz. (8 per case)
    21 lbs.
    Log in for availability
    Log in for availability
    Case of 8 (1/Itm/Bx)
    Case of 8 (1/Itm/Bx) - $711.44
    $711.44
    Case of 8 (1/Itm/Bx)
       $711.44
    Supplier Number: BIT-100-DOMESTIC
  • SteraMist BIT Solution for use with all SteraMist Systems
    15x10x14"
    1 gal. (4 per case)
    36 lbs.
    Log in for availability
    Log in for availability
    Case of 1 (1/Itm/Bx)
    Case of 1 (1/Itm/Bx) - $1,508.08
    $1,508.08
    Case of 1 (1/Itm/Bx)
       $1,508.08
    Supplier Number: BIT-400-DOMESTIC
  • SteraMist BIT Solution for use with all SteraMist Systems
    19x13x16"
    10 L (2 per case)
    45 lbs.
    Log in for availability
    Log in for availability
    Each(10l)
    Each(10l) - $2,251.81
    $2,251.81
    Each(10l)
       $2,251.81
    Supplier Number: BIT-400-10L
  • SteraMist BIT Solution for use with all SteraMist Systems
    12x11x18"
    5 gal. container (1 per case)
    45 lbs.
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $2,892.32
    $2,892.32
    Each(1/Itm/Bx)
       $2,892.32
    Supplier Number: BIT-400-5G
  • SteraMist BIT Solution for use with all SteraMist Systems
    --
    55 gal.
    550 lbs.
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $15,582.51
    $15,582.51
    Each(1/Itm/Bx)
       $15,582.51
    Supplier Number: BIT-40055GL