SteraMist Environment System
SteraMist Environment System - 110/120 V includes three remote applicators, three stands and mounts, one tool kit, one DI water cartridge
  76919-446
 :  ENV-303-110
SteraMist Environment System
  76919-446
SteraMist Environment System
SteraMist Environment System
SteraMist Environment System - 110/120 V includes three remote applicators, three stands and mounts, one tool kit, one DI water cartridge
$148,018.85
Each
The SteraMist Environment System, the most intelligent mobile unit, delivers disinfection and decontamination with unparalleled scalability.
  • 3 removable 30’ cords and applicator stands
  • Intuitive HMI and custom cycle creation to meet all GMP requirements
  • Features remote operation and hand-held modes
  • Contains applicators able to treat areas of any size
Ideal for outbreaks and severe decontamination and deodorizing needs.
Delivery information: Includes 3× remote applicators (single point connectors with 30’ cords), 3× remote applicator stands and mounts, tool kit, 1 gal. DI water cartridge.
  • Désignation:
    SteraMist Environment System - 110/120 V includes three remote applicators, three stands and mounts, one tool kit, one DI water cartridge
Services
  •  
    Description
  • 76919-422
    Repair work to maintain your SteraMist equipment, quoted at time of receipt of unit and after evaluation
    Each - $1,611.60
  • 76919-416
    Additional one-year warranty for purchased SteraMist products and accessories
    Each - $5,640.60
  • 76919-432
    Training and continued support on SteraMist equipment, mandatory for first time users (i.e. protocol and policy development, troubleshooting, assistance as-needed etc.)
    Each - $10,475.40
  • 76919-430
    Corrective calibration to ensure the SteraMist Environment System is running on proper levels, anually recommended
    Each - $1,087.83
Accessories
  •  
    Description
  • 76919-436
    Replacement SteraMist applicator, for SteraMist Surface Unit and SteraMist Environment System
    Each - $8,676.96
  •  
    Description
  • 76919-444
    Stainless steel cart (only)
    Each - $2,697.99