ContactUsLinkComponent
PHCbi VIP® ECO SMART Ultra-Low Temperature Upright Freezers –80 °C
PHCbi VIP® ECO SMART Ultra-Low Temperature Upright Freezers –80 °C
Supplier:  PHC Corporation
Certificates
undefined
PHCbi VIP® ECO SMART Ultra-Low Temperature Upright Freezers –80 °C
Supplier:  PHC Corporation
Specifications
  • Catalog No:
  • CA76637-152
  • CA76637-148
  • Defrost:
  • Variable
  • Variable
  • Electrical:
  • 115 V
  • 115 V
  • Shelves/drawers:
  • 3 adjustable shelves
  • 3 adjustable shelves
  • Door type:
  • 2 insulated stainless steel inner doors, 1 external door
  • 2 insulated stainless steel inner doors, 1 external door