Masterflex® USB A Serial Adapter Cables, Avantor® | Avantor
Masterflex® USB A Serial Adapter Cables, Avantor®
Masterflex® USB A Serial Adapter Cables, Avantor®
Certificates
Masterflex® USB A Serial Adapter Cables, Avantor®
2 Options Available Below
Product Details & Documents
Optimize performance of your Masterflex® MasterSense™ or Ismatec® Reglo Touchscreen pump.
  • Connect your drive via USB A for serial control
Not your standard USB cable. Use one of these cables to connect a Masterflex® MasterSense™ and Ismatec® Reglo Touchscreen pumps to a PC or PLC for serial (RS232) communicaiton. Cables features special capability to support serial interface communication. Chose USB A to USB A for communication to computers. The three wire cable is desinged for serial communcation with PLC applications.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • MFLX78006-90
    Serial coms cable USB-A - USB-A, 8'
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $157.00
    $157.00
    Each
       $157.00
    Supplier Number: 78006-90
  • MFLX78006-91
    Serial Coms Cable: USB-A to PLC (3 wire), 8 ft.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $157.00
    $157.00
    Each
       $157.00
    Supplier Number: 78006-91