Adjustable Sampling Tubes for Bioreactors, M10
Supplier: Chemglass
Jump to:
À propos de cet article
These sampling tubes are adjustable to permit the culture fluid to be sampled at any desired level.
Tubing is ¹/₄" OD. Stainless steel components manufactured from 316 L with an electropolished surface finish of 20Ra or better.
3 Options Available Below
Product Details & Documents
These sampling tubes are adjustable to permit the culture fluid to be sampled at any desired level.
Tubing is ¹/₄" OD. Stainless steel components manufactured from 316 L with an electropolished surface finish of 20Ra or better.
Product Family Options
Informations sur le produit
- LongueurVolumeDésignationAvailabilityPrice
- Supplier Number: CLS-1380-21
- Supplier Number: CLS-1383-08
- Supplier Number: CLS-1385-13