Accessories for Identifier System
Sampling swab, Dimensions: 110×14×5 mm (4.3×0.55×0.2")
Catalog # 76848-020
Supplier: STAT PEEL INC
Fournisseur #: C00553
Jump to:
Product Details & Documents
Spécifications
- Désignation:Sampling swab, Dimensions: 110×14×5 mm (4.3×0.55×0.2")