Filtration Slides for Identifier System
À propos de cet article
The nanoporous filtration membrane inside the Stat Peel filtration slide allows the collection of aerosol particles down to the nanometer range.
- Proprietary, inert nanoporous filtration membrane for aerosol particle collection
- 1 mm² gold-coated filter surface
- Aluminum filtration slide body
- Internal flow channels
- QR code for traceability
The Stat Peel filtration slide was designed to operate together with the air sampling badge to comply with the particle size fractions defined in ISO 7708. The nanoporous filtration membranes retain particles on a gold-coated, inert surface for inspection later by the Stat Peel Identifier Raman system.
Filtration slides with different internal flow channels separate the collected particles into size bins for a size dependent inspection. The small, flat, and gold-coated filtration membrane is also an ideal substrate to inspect collected particles by SEM. The slides are available in boxes of 90 pieces for insertion into the Identifier System or in smaller boxes of 20 pieces for use with the sampling kit.
: In compliance with ISO 7708.
8 Options Available Below
Product Details & Documents
The nanoporous filtration membrane inside the Stat Peel filtration slide allows the collection of aerosol particles down to the nanometer range.
- Proprietary, inert nanoporous filtration membrane for aerosol particle collection
- 1 mm² gold-coated filter surface
- Aluminum filtration slide body
- Internal flow channels
- QR code for traceability
The Stat Peel filtration slide was designed to operate together with the air sampling badge to comply with the particle size fractions defined in ISO 7708. The nanoporous filtration membranes retain particles on a gold-coated, inert surface for inspection later by the Stat Peel Identifier Raman system.
Filtration slides with different internal flow channels separate the collected particles into size bins for a size dependent inspection. The small, flat, and gold-coated filtration membrane is also an ideal substrate to inspect collected particles by SEM. The slides are available in boxes of 90 pieces for insertion into the Identifier System or in smaller boxes of 20 pieces for use with the sampling kit.
: In compliance with ISO 7708.
Product Family Options
Informations sur le produit
- DésignationDimensionsDetailsAvailabilityPrice
- Filtration slides, inhalable, 1 channel185×112×84 mm (7.3×4.4×3.3")1.8 kg (4 lbs.)Log in pour la disponibilitéBox of 90$2,272.43Supplier Number: A00552
- Filtration slides, respirable, 3 channels185×112×84 mm (7.3×4.4×3.3")1.8 kg (4 lbs.)Log in pour la disponibilitéBox of 90$3,787.39Supplier Number: A00551
- Filtration slides, respirable, 1 channel185×112×84 mm (7.3×4.4×3.3")1.8 kg (4 lbs.)Log in pour la disponibilitéBox of 90$2,272.43Supplier Number: A00471
- Filtration slides, respirable, 2 channels185×112×84 mm (7.3×4.4×3.3")1.8 kg (4 lbs.)Log in pour la disponibilitéBox of 90$3,029.91Supplier Number: A00550
- Filtration slides, respirable, 1 channel94×84×33 mm (3.7×3.3×1.3")355 g (12.5 oz.)Log in pour la disponibilitéBox of 20$504.99Supplier Number: A00059
- Filtration slides, respirable, 2 channels94×84×33 mm (3.7×3.3×1.3")355 g (12.5 oz.)Log in pour la disponibilitéBox of 20$701.04Supplier Number: A00060
- Filtration slides, respirable, 3 channels94×84×33 mm (3.7×3.3×1.3")355 g (12.5 oz.)Log in pour la disponibilitéBox of 20$841.65Supplier Number: A00061
- Filtration slides, inhalable, 1 channel94×84×33 mm (3.7×3.3×1.3")355 g (12.5 oz.)Log in pour la disponibilitéBox of 20$504.99Supplier Number: A00473