ContactUsLinkComponent
miniPCR® DNA Fingerprinting Lab: Shark Attack!
miniPCR® DNA Fingerprinting Lab: Shark Attack!
Catalog # 470352-008
Supplier:  AMPLYUS LLC
undefined
miniPCR® DNA Fingerprinting Lab: Shark Attack!
Catalog # 470352-008
Supplier:  AMPLYUS LLC
Supplier Number:  KT-1500-03
Specifications
  • Description:
    miniPCR® DNA fingerprinting: shark attack lab
  • No. of Lab Groups:
    8
  • Cat. no.:
    470352-008
Specifications
About this item
Use gel electrophoresis to compare DNA fingerprints from the shark attacks and determine if a single shark is to blame.
  • Techniques: Micropipetting, gel electrophoresis
  • Topics: Forensics, genetic polymorphism, environmental science, biotechnology
  • Time required: Can be completed in one 45-minute class period
  • Level: Advanced high school through general high school
Help marine biologists understand the source of shark attacks that are frightening Australian beachgoers! This guided-inquiry activity exposes students to the fundamental principles of DNA fingerprinting, which is routinely used to identify individuals based on their genetic makeup. DNA fingerprinting has a broad range of human applications, including forensics and paternity testing, but it is also an important tool in ecology. Students will determine if a single shark has attacked multiple times or if there are several different attackers.
Reagents and consumables supplied by user:
- Agarose (electrophoresis grade)
- DNA stain (e.g., GelGreen™)
- Gel electrophoresis buffer (e.g., 1X TBE)
- Distilled or deionized H₂O (to dilute buffer concentrate – not included in Lab Companion Kit)
- Microtubes
Required equipment:
- Micropipettes and tips
- Gel electrophoresis and fluorescence visualization system
Storage notes:
- Reagents require freezer storage
- Reagents are stable for 12 months when stored in the freezer
Accessories information: The learning lab companion kit (catalog no. 470352-026) contains all the consumables (except water) needed for a class of 32 students. These reagents and consumables can also be sourced independently.
Delivery: The DNA Fingerprinting Shark Attack Lab kit contains reagents for 8 lab groups: 4 DNA samples for fingerprinting analysis; 1 tube of DNA molecular weight marker (DNA Ladder); 6X gel loading dye. Note: Reagents are compatible with blueGel™ or other standard horizontal electrophoresis systems.