ContactUsLinkComponent
MULTISCREEN™ Mouse CB2 CHO-K1 β-Arrestin2 HTS Membranes
undefined
MULTISCREEN™ Mouse CB2 CHO-K1 β-Arrestin2 HTS Membranes
  • Storage conditions:
    LN₂ (Below –135 ℃)
Frequently Bought Together