MULTISCREEN™ Human ZnT8 CHO-K1 HTS Membranes
Fournisseur: MULTISPAN INC
Certificats
À propos de cet article
Spécifications
- Storage conditions:LN₂ (Below –135 ℃)
Spécifications
Frequently Bought Together
VWR® Multipurpose Containers, HDPE
Fournisseur: VWR International
Histoplex™ Histology Containers, Starplex®
Fournisseur: Starplex
Acide chlorhydrique 1.0 N analytical reagent, VWR Chemicals BDH®
Fournisseur: VWR International
Acétone ≥99.5% ACS
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
VWR® Sleeve Stoppers
Fournisseur: VWR International
Masterflex® Hose Clamps, Acetal Copolymer, Avantor®
Fournisseur: VWR International
Glucose Tolerance Drinks, Azer Scientific
Fournisseur: Azer Scientific
A.P. Green Fire Assay Crucibles, HarbisonWalker
Fournisseur: HarbisonWalker
Accessories for Animal Tattoo Pigments, Needles, and Supplies, AIMS
Fournisseur: Aims
DpnI, Restriction Enzymes, New England Biolabs
Fournisseur: New England Biolabs (NEB)
Nitric acid 65%, Suprapur® for trace metal analysis, Supelco®
Fournisseur: MilliporeSigma
Puritan PurFlock Ultra® Flocked Swabs, Puritan Medical Products
Fournisseur: Puritan Medical Products
Buffer, EDTA Buffer Solution pH 8,0 (0,5 mol/L), ROCKLAND
Fournisseur: Rockland Immunochemical
Bovine Serum Albumin (BSA)
Fournisseur: Rockland Immunochemical
OSOM® Ultra Strep A Test Kits, Sekisui Diagnostics
Fournisseur: Sekisui Diagnostics