MULTISCREEN™ Human SST5 CHO-K1 Gα16 HTS Membranes
Supplier: MULTISPAN INC
Certificates
About this item
Specifications
- Storage conditions:LN₂ (Below –135 ℃)
Specifications
Frequently Bought Together
VWR® Traceable® Conductivity Calibration Standards (CRM)
Supplier: VWR International
Dulbecco's phosphate-buffered saline (DPBS), Mediatech
Supplier: Corning
Hydrogen peroxide 3% (w/w) in aqueous solution stabilized, VWR Chemicals BDH®
Supplier: VWR International
VWR® Specimen Containers with Dual Click
Supplier: VWR International
VWR® Sleeve Stoppers
Supplier: VWR International
VWR® Ultra Pure Grade Phosphate Buffered Saline
Supplier: VWR
Gauze Pads, DUKAL™ Corporation
Supplier: Dukal
Single-Use Needles, BD Medical
Supplier: BD
Water, distilled
Supplier: GROWCELLS
Orion™ pH Buffer Individual Use Pouches, Thermo Scientific
Supplier: Thermo Fisher Scientific
Alcool ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
Lithium bromide ≥99.0% (by titrimetric analysis)
Supplier: TCI America
D-(+)-Raffinose pentahydrate ≥98.0% (by HPLC)
Supplier: TCI America
Diversey™ Oxivir® Tb Disinfectant Solution, Antylia Scientific
Supplier: Antyila Scientific
SUBSTRATE TMB MEMBRANE PEROXIDASE 1 L
Supplier: Rockland Immunochemical