ContactUsLinkComponent
MULTISCREEN™ Rat EP2 HEK293T HTS Membranes
MULTISCREEN™ Rat EP2 HEK293T HTS Membranes
Supplier:  MULTISPAN INC
Certificates
undefined
MULTISCREEN™ Rat EP2 HEK293T HTS Membranes
Supplier:  MULTISPAN INC
Specifications
  • Storage conditions:
    LN₂ (Below –135 ℃)