MULTISCREEN™ Human EBI2 CHO-K1 Gαqi5 HTS Membranes
Supplier: MULTISPAN INC
Certificates
About this item
Specifications
- Storage conditions:LN₂ (Below –135 ℃)
Specifications
Frequently Bought Together
Preserved Starfish
Supplier: Avantor
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Nunc-Immuno™ MicroWell™ 96-Well Plates, Thermo Scientific
Supplier: Thermo Fisher Scientific
Sodium hydroxide 0.100 N (N/10) in aqueous solution
Supplier: Ricca Chemical
Toluène ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Lithium Chloride
Supplier: Ward's Science
SYRINGE WITH NEEDLE 5ML
Supplier: AMX INDUSTRIAL TRADE MX
HyFlex® 11-318 Touch Screen-Capable Gloves, Uncoated, Ansell
Supplier: Ansell Canada
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.0-101.0% USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
TelN Protelomerase
Supplier: New England Biolabs (NEB)
VACUUBRAND® ME 1C Oil-Free Chemistry Diaphragm Pump, Trade-Up Promotion
Supplier: Brandtech