MULTISCREEN™ Rat GPR88 CHO-K1 HTS Membranes
- Storage conditions:LN₂ (Below –135 ℃)
Frequently Bought Together
Leakbuster™ Specimen Containers
: Starplex
VWR® Sleeve Stoppers
: VWR International
Méthanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
: MilliporeSigma
TETRAHYDROFURAN OMNISOLV 4L
: MilliporeSigma
Orion™ ROSS Ultra™ Low Maintenance pH/ATC Triode™ Combination Electrodes, Thermo Scientific
: Thermo Fisher Scientific
DNA Clean and Concentrator™ Kits, Zymo Research
: Zymo Research
Éthanol dénaturé, anhydrous
: AVANTOR PERFORMANCE MATERIAL LLC
Formalin, VWRClick, neutral buffered prefill
: VWR International
Calcium chloride 2.75% (w/v) in aqueous solution, VWR Chemicals BDH®
: VWR International
Orion™ pH Buffer Bottles, Thermo Scientific
: Thermo Fisher Scientific
Propanol-2 ≥70%, VWR Chemicals BDH®
: VWR International
Pharmacy Bags, Reclosable, Elkay Plastics
: Elkay Plastics
Sodium peroxide, granular for analysis, Supelco®
: MilliporeSigma
Polysorbate 80 NF, Multi-Compendial, J.T.Baker®
: AVANTOR PERFORMANCE MATERIAL LLC