MULTISCREEN™ Human mGluR7 HEK293T Gαqi5 HTS Membranes
Supplier: MULTISPAN INC
Certificates
About this item
Specifications
- Storage conditions:LN₂ (Below –135 ℃)
Specifications
Frequently Bought Together
INCYTO C-Chip™ Disposable Hemacytometers, SKC
Supplier: MEDONE SURGICAL, INC
BD GasPak™ EZ Container Systems, BD Diagnostics
Supplier: BD
Optical Lens Cleaner, FoxWest Sales
Supplier: Decon Labs
VWR® Sleeve Stoppers
Supplier: VWR International
CHEMets® Visual Kit, Hydrogen Peroxide Test Kit, CHEMetrics
Supplier: CHEMetrics
VWR® Blood Bank Saline Solutions
Supplier: VWR International
VWR® Multipurpose Containers, HDPE
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Multistage Gas Regulators with Stainless Steel Diaphragms
Supplier: VWR International
Tryptone to 2xYT Broth, Life Technologies
Supplier: BD
Single-Use Needles, BD Medical
Supplier: BD
VACUETTE® Secondary Tubes, Greiner Bio-One
Supplier: Greiner Bio-One
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
MEDIUM SANITIZER NEUZ AGAR LEC ST PK100
Supplier: BD
Corning® rat collagen I (from tail tendon)
Supplier: Corning
Solvent Resistant Pen, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Columbia Agar, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
Acetone ACS
Supplier: Ricca Chemical
Sulfuric acid 0.1 N in aqueous solution, BAKER ANALYZED® volumetric solution, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC