MULTISCREEN™ Rat mGluR4 HEK293T HTS Membranes
Fournisseur: MULTISPAN INC
Certificats
À propos de cet article
Spécifications
- Storage conditions:LN₂ (Below –135 ℃)
Spécifications
Frequently Bought Together
Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
Fournisseur: Thermo Scientific
Single-Use Needles, BD Medical
Fournisseur: BD
Acide chlorhydrique 0.10 N analytical reagent, VWR Chemicals BDH®
Fournisseur: VWR International
Orion™ pH Buffer Bottles, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Sanicult™ Hygiene Monitoring Test Kits, Starplex®
Fournisseur: Starplex
Acétonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Fournisseur: MilliporeSigma
NORM-JECT® Luer Lock Sterile Syringes, Air-Tite Products
Fournisseur: Air-Tite
BD GasPak™ EZ Container Systems, BD Diagnostics
Fournisseur: BD
pH Reference Standard Buffers, VWR Chemicals BDH®
Fournisseur: BDH
Ocean Pacific® Ninja™ Powder-Free Nitrile Medical Examination Gloves, Medicom
Fournisseur: Medicom
Nitric acid 67 - 70%, OmniTrace® for trace metal analysis, Supelco®
Fournisseur: MilliporeSigma
Ward's® Live <i>Lactobacillus delbrueckii ss. bulgaricus</i> Culture
Fournisseur: Avantor
Circular Cover Glass, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
2,2,4-Triméthylpentane, Accusolv™, distilled
Fournisseur: Anachemia
Pierce™ Blocker™ Casein in PBS, Thermo Scientific
Fournisseur: Thermo Scientific
Ether diéthylique ≥98% stabilised ACS, VWR Chemicals BDH®
Fournisseur: VWR International