MULTISCREEN™ Mouse H4 HEK293T Gαqi5 HTS Membranes
- Storage conditions:LN₂ (Below –135 ℃)
Frequently Bought Together
Phenols, CHEMets Visual Kit, CHEMetrics
: CHEMetrics
Polysorbate 80 solution analytical reagent, VWR Chemicals BDH®
: VWR International
SporAmpule® Liquid Biological Indicator
: HARDY DIAGNOSTICS CA
Solvent Resistant Pen, Electron Microscopy Sciences
: Electron Microscopy Sciences
Chloroform ≥99.9% stabilized, OmniSolv®, Supelco®
: MilliporeSigma
Sodium dodecyl sulfate (SDS) ≥99.0%, OmniPur®, Millipore®
: MilliporeSigma
Hydrogen peroxide 30% stabilized, CMOS for microelectronic use, J.T.Baker®
: AVANTOR PERFORMANCE MATERIAL LLC