MULTISCREEN™ Ferret GIP HEK293T HTS Membranes
Fournisseur: MULTISPAN INC
Certificats
À propos de cet article
Spécifications
- Storage conditions:LN₂ (Below –135 ℃)
Spécifications
Frequently Bought Together
n-Hexane ≥95% (by GC, corrected for water content) for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
2,2,4-Triméthylpentane ≥99.0% ACS
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
VWR® Self-Seal Sterilization Pouches
Fournisseur: VWR International
VWR® Traceable® Conductivity Calibration Standards (CRM)
Fournisseur: VWR International
Acide chlorhydrique 3.0 N analytical reagent, VWR Chemicals BDH®
Fournisseur: VWR International
Méthanol ≥99.8% ACS for HPLC, Supelco®
Fournisseur: MilliporeSigma
VWR® Sleeve Stoppers
Fournisseur: VWR International
NORM-JECT® Luer-Lock and Luer-Slip Bulk Syringes, Two-Part, Nonsterile, Air-Tite
Fournisseur: Air-Tite
Boron trifluoride diethyl ether complex (≥46.5% BF₃)
Fournisseur: Thermo Scientific Chemicals
tri-Sodium citrate dihydrate ≥99% (by non aqueous titration), granular, BAKER ANALYZED® ACS, J.T.Baker®
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
SYRINGE WITH NEEDLE 5ML
Fournisseur: AMX INDUSTRIAL TRADE MX
Corning® Ham's F-12 Medium, Corning
Fournisseur: Corning
VWR® Single-Edged Razor Blades
Fournisseur: VWR International
Single-Use Needles, BD Medical
Fournisseur: BD