MULTISCREEN™ Human GPR43 HEK293T HTS Membranes
- Storage conditions:LN₂ (Below –135 ℃)
Frequently Bought Together
VWR® Sleeve Stoppers
: VWR International
VWR® Razor Blades
: VWR International
Acetonitrile ≥99.9%, OmniSolv® LC-MS for LC-MS, Supelco®
: MilliporeSigma
Méthanol, anhydrous ≥99.8% (by GC), GR ACS ACS, Supelco®
: MilliporeSigma
High Sensitivity RNA ScreenTape Analysis
: AGILENT TECHNOLOGIES (GENOMICS) CA
Corning® Trypsin
: Corning