MULTISCREEN™ Rat GPR41 HEK293T HTS Membranes
- Storage conditions:LN₂ (Below –135 ℃)
Frequently Bought Together
VWR® Multistage Gas Regulators with Stainless Steel Diaphragms
: VWR International
MQuant® pH Test Strips, Supelco®,MilliporeSigma
: MilliporeSigma
Propanol-2 ≥70%, VWR Chemicals BDH®
: VWR International
Leakbuster™ Specimen Containers
: Starplex
VWR® Sleeve Stoppers
: VWR International
Méthanol ≥99.8% ACS, VWR Chemicals BDH®
: VWR International
Méthanol ≥99.9%, OmniSolv® for purge & trap, Supelco®
: MilliporeSigma
B352 Specimen Bottles and Caps, Simport Scientific
: Simport Scientific
Phalloidin Conjugates, Biotium
: Biotium
VWR® Single-Edged Razor Blades
: VWR International