MULTISCREEN™ Human CXCR1 HEK293T HTS Membranes
Fournisseur: MULTISPAN INC
Certificats
À propos de cet article
Spécifications
- Storage conditions:LN₂ (Below –135 ℃)
Spécifications
Frequently Bought Together
Leakbuster™ Specimen Containers
Fournisseur: Starplex
VWR® Sleeve Stoppers
Fournisseur: VWR International
VWR® TBE 10X Liquid Concentrate, Ultra Pure Grade
Fournisseur: VWR
B352 Specimen Bottles and Caps, Simport Scientific
Fournisseur: Simport Scientific
Histoplex™ Histology Containers, Starplex®
Fournisseur: Starplex
pH Reference Standard Buffers, VWR Chemicals BDH®
Fournisseur: BDH
Acide chlorhydrique 0.5 N analytical reagent, VWR Chemicals BDH®
Fournisseur: VWR International
Air-Tite Vet Premium Hypodermic Needles, Air-Tite Products
Fournisseur: Air-Tite
eShield™ Sterile Cell Phone Cover, Whitney Medical Solutions
Fournisseur: Heathrow Scientific
BathBrite™ Cleaner
Fournisseur: Decon Labs
VWR® Single-Edged Razor Blades
Fournisseur: VWR International
Formaldehyde (Paraformaldehyde) 16% in aqueous solution, EM Grade
Fournisseur: Electron Microscopy Science
OSOM® Ultra Strep A Test Kits, Sekisui Diagnostics
Fournisseur: Sekisui Diagnostics
Corning® Ham's F-12 Medium, Corning
Fournisseur: Corning
Columbia Agar, Hardy Diagnostics
Fournisseur: HARDY DIAGNOSTICS CA