MULTISCREEN™ Mouse CCR6 HEK293T HTS Membranes
Supplier: MULTISPAN INC
Certificates
About this item
Specifications
- Storage conditions:LN₂ (Below –135 ℃)
Specifications
Frequently Bought Together
Sodium thiosulfate 0.100 N (N/10) in aqueous solution
Supplier: Ricca Chemical
Accessories for Advanced® Multi-Sample Osmometer, Model 2020, Advanced Instruments
Supplier: Advanced Instruments
Propanol-2 ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Sleeve Stoppers
Supplier: VWR International
Air-Tite Vet Premium Hypodermic Needles, Air-Tite Products
Supplier: Air-Tite
Acetonitrile ≥99.9%, OmniSolv® LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
Orion™ pH Electrode Cleaning and Storage Solutions, Thermo Scientific
Supplier: Thermo Fisher Scientific
PREempt® Concentrate Disinfectants, Contec
Supplier: Contec
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Acide chlorhydrique 6.0 N analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
Corning® PBS (Phosphate-Buffered Saline), Corning
Supplier: Corning
Formaldehyde (Paraformaldehyde) 16% in aqueous solution, EM Grade
Supplier: Electron Microscopy Science
Circular Cover Glass, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
qScript™ cDNA SuperMix, QuantaBio
Supplier: Quantabio
SELF CONTAINED BIO INDICATOR
Supplier: Steris
Corning® Matrigel® Basement Membrane Matrix, Corning
Supplier: Corning