MULTISCREEN™ Mouse CB1 CHO-K1 HTS Membranes
MULTISCREEN™ Mouse CB1 CHO-K1 HTS Membranes
MULTISCREEN™ Mouse CB1 CHO-K1 HTS Membranes
MULTISCREEN™ Mouse CB1 CHO-K1 HTS Membranes
  • Catalog No:
  • 76922-294
  • 76920-712
  • 76920-366
  • 76923-188
  • 76921-776
  • 76922-746
  • 76921-676
  • 76921-980
  • Storage conditions:
  • LN₂ (Below –135 ℃)
  • LN₂ (Below –135 ℃)
  • LN₂ (Below –135 ℃)
  • LN₂ (Below –135 ℃)
  • LN₂ (Below –135 ℃)
  • LN₂ (Below –135 ℃)
  • LN₂ (Below –135 ℃)
  • LN₂ (Below –135 ℃)
Product Family Options
  • Désignation
    Format de cellule
    Donneur unique/donneurs regroupés
    Taille
  • high receptor expression
    Cryopreserved
    Single donor
    100 mg
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $51,571.67
    $51,571.67
    Each(1/Itm/Bx)
       $51,571.67
     : MCM12291A100MG
    Specifications
    Cat. No.
    76922-294
    Storage conditions
    LN₂ (Below –135 ℃)
  • high receptor expression
    Cryopreserved
    Single donor
    10 mg
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $7,368.47
    $7,368.47
    Each(1/Itm/Bx)
       $7,368.47
     : MCM1229-1A10MG
    Specifications
    Cat. No.
    76920-712
    Storage conditions
    LN₂ (Below –135 ℃)
  • high receptor expression
    Cryopreserved
    Single donor
    1 mg
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $1,430.69
    $1,430.69
    Each(1/Itm/Bx)
       $1,430.69
     : MCM1229-1A-1MG
    Specifications
    Cat. No.
    76920-366
    Storage conditions
    LN₂ (Below –135 ℃)
  • high receptor expression
    Cryopreserved
    Single donor
    5 mg
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $5,157.93
    $5,157.93
    Each(1/Itm/Bx)
       $5,157.93
     : MCM1229-1A-5MG
    Specifications
    Cat. No.
    76923-188
    Storage conditions
    LN₂ (Below –135 ℃)
  • low receptor expression
    Cryopreserved
    Single donor
    100 mg
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $51,571.67
    $51,571.67
    Each(1/Itm/Bx)
       $51,571.67
     : MCM12291B100MG
    Specifications
    Cat. No.
    76921-776
    Storage conditions
    LN₂ (Below –135 ℃)
  • low receptor expression
    Cryopreserved
    Single donor
    10 mg
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $7,368.47
    $7,368.47
    Each(1/Itm/Bx)
       $7,368.47
     : MCM1229-1B10MG
    Specifications
    Cat. No.
    76922-746
    Storage conditions
    LN₂ (Below –135 ℃)
  • low receptor expression
    Cryopreserved
    Single donor
    1 mg
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $1,430.69
    $1,430.69
    Each(1/Itm/Bx)
       $1,430.69
     : MCM1229-1B-1MG
    Specifications
    Cat. No.
    76921-676
    Storage conditions
    LN₂ (Below –135 ℃)
  • low receptor expression
    Cryopreserved
    Single donor
    5 mg
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $5,157.93
    $5,157.93
    Each(1/Itm/Bx)
       $5,157.93
     : MCM1229-1B-5MG
    Specifications
    Cat. No.
    76921-980
    Storage conditions
    LN₂ (Below –135 ℃)