MULTISCREEN™ Rat B2 HEK293T HTS Membranes
Fournisseur: MULTISPAN INC
Certificats
À propos de cet article
Spécifications
- Storage conditions:LN₂ (Below –135 ℃)
Spécifications
Frequently Bought Together
Acide sulfurique 0.02 N in aqueous solution analytical reagent, VWR Chemicals BDH®
Fournisseur: VWR International
Air-Tite Vet Premium Hypodermic Needles, Air-Tite Products
Fournisseur: Air-Tite
B352 Specimen Bottles and Caps, Simport Scientific
Fournisseur: Simport Scientific
pH Reference Standard Buffers, VWR Chemicals BDH®
Fournisseur: BDH
BD Vacutainer® Venous Blood Collection Tubes
Fournisseur: BD
SYRINGE FOR INSULIN WITH NEEDLE 1ML
Fournisseur: AMX INDUSTRIAL TRADE MX
Sodium sulfate, anhydrous ≥99.0%, granular, Tracepur® 10-60 mesh, Supelco®
Fournisseur: MilliporeSigma
Méthanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Fournisseur: MilliporeSigma
INCYTO C-Chip™ Disposable Hemacytometers, SKC
Fournisseur: MEDONE SURGICAL, INC
Single-Use Needles, BD Medical
Fournisseur: BD
Méthanol ≥99.9%, HiPerSolv CHROMANORM® for HPLC-MS, suitable for LC-MS, UPLC, UHPLC, Ultra HPLC, VWR Chemicals BDH®
Fournisseur: VWR International
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Fournisseur: MilliporeSigma
Bromocresol green 0.1% (w/v) in aqueous solution indicator
Fournisseur: Ricca Chemical
High Sensitivity RNA ScreenTape Analysis
Fournisseur: AGILENT TECHNOLOGIES (GENOMICS) CA
Insulin Syringes with Permanently Attached Needles, BD Medical
Fournisseur: Cardinal Health
Ethyl acetate ≥99.8% for HPLC, Supelco®
Fournisseur: MilliporeSigma
Accessories for HemataSTAT® II Centrifuge, Separation Technology
Fournisseur: Separation Technology
STORAGE SOLUTION PH ELECTRODE
Fournisseur: Oakton