MULTISCREEN™ Human Beta2 CHO-K1 Gαqi5 HTS Membranes
- Storage conditions:LN₂ (Below –135 ℃)
Frequently Bought Together
Méthanol ≥99.8% ACS, VWR Chemicals BDH®
: VWR International
VWR® Sleeve Stoppers
: VWR International
Acetonitrile ≥99.9%, OmniSolv® LC-MS for LC-MS, Supelco®
: MilliporeSigma
Corning® RPMI 1640, Corning
: Corning
VWR® Disposable Petri Dishes
: VWR International
2-Propanol ≥99.8% for HPLC, Supelco®
: MilliporeSigma
B352 Specimen Bottles and Caps, Simport Scientific
: Simport Scientific
Advanced® Bilirubin STAT-Analyzer™, Model BR2, Advanced Instruments
: Advanced Instruments