MULTISCREEN™ Mouse A2A HEK293T HTS Membranes
Supplier: MULTISPAN INC
Certificates
About this item
Specifications
- Storage conditions:LN₂ (Below –135 ℃)
Specifications
Frequently Bought Together
RNase AWAY® and DNA AWAY™ Surface Decontaminants, Molecular BioProducts
Supplier: Thermo Scientific
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
SYRINGE STERILE WITHOUT NEEDLE 10ML
Supplier: AMX INDUSTRIAL TRADE MX
Acide sulfurique 1.0 N in aqueous solution analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
Eveready® Alkaline Batteries, Bulbtronics
Supplier: Bulbtronics
Technical Buffer Solutions, Sachets
Supplier: Mettler Toledo
TOTAL CHLORINE IND SOL 473ML
Supplier: Hach
DAPT ≥98% (by NMR)
Supplier: Enzo Life Sciences
Accessories for perfectION™ Combined Ion Selective Electrode electrodes
Supplier: Mettler Toledo
AGAR BRAIN HEART INFUSION 450 G
Supplier: BECTON DICKINSON DE MEXICO, S. MX
Sanicult™ Hygiene Monitoring Test Kits, Starplex®
Supplier: Starplex
LabKlenz™ 100 Alkaline Detergents
Supplier: Steris
Sol-M® Standard Insulin Syringes
Supplier: SOL-MILLENNIUM CANADA, INC. CA
Difco™ Raka-Ray No.3 Medium, BD
Supplier: BD
Skim Milk Powder, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA